3-(4-Fluoro-2-methoxyphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

458769-5G 458769-1G 458769-25G 458769-500MG
77629-480EA 1484.23 USD
77629-480 77638-920 77659-448 77650-930
3-(4-Fluoro-2-methoxyphenyl)oxetan-3-ol ≥95%
3-(4-Fluoro-2-methoxyphenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 198.1909 g/mol
MW: C₁₀H₁₁FO₃
Storage Temperature: Ambient
CAS Number: 3037703-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR