2-(5-Chloro-2-fluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

457604-5G 457604-1G 457604-500MG
77629-476EA 1484.23 USD
77629-476 77638-914 77650-926
2-(5-Chloro-2-fluorophenyl)propan-2-ol ≥95%
2-(5-Chloro-2-fluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₉H₁₀ClFO
Storage Temperature: Ambient
CAS Number: 1506675-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR