2-(5-Bromobenzo[d][1,3]dioxol-4-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

457589-1G 457589-5G 457589-500MG
77638-910EA 566 USD
77638-910 77659-442 77650-920
2-(5-Bromobenzo[d][1,3]dioxol-4-yl)propan-2-ol ≥95%
2-(5-Bromobenzo[d][1,3]dioxol-4-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 259.097 g/mol
MW: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
CAS Number: 249636-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR