2,4-Difluoro-5-isopropoxy-3-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

457532-5G 457532-25G 457532-1G 457532-500MG
77629-472EA 1484.23 USD
77629-472 77655-852 77642-952 77650-914
2,4-Difluoro-5-isopropoxy-3-methoxybenzaldehyde ≥95%
2,4-Difluoro-5-isopropoxy-3-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: 230.208 g/mol
MW: C₁₁H₁₂F₂O₃
Storage Temperature: Ambient
CAS Number: 3037703-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR