1-(6-Bromo-2,3-difluorophenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

457120-5G 457120-1G 457120-25G 457120-500MG
77629-466EA 1484.23 USD
77629-466 77638-900 77655-850 77646-930
1-(6-Bromo-2,3-difluorophenyl)prop-2-en-1-ol ≥95%
1-(6-Bromo-2,3-difluorophenyl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 249.052 g/mol
MW: C₉H₇BrF₂O
Storage Temperature: Ambient
CAS Number: 3037693-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR