(3-Ethoxy-2,6-difluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

457105-5G 457105-25G 457105-1G 457105-500MG
77635-030EA 328 USD
77635-030 77659-440 77642-950 77646-928
(3-Ethoxy-2,6-difluorophenyl)trimethylsilane ≥95%
(3-Ethoxy-2,6-difluorophenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 230.326 g/mol
MW: C₁₁H₁₆F₂OSi
Storage Temperature: Ambient
CAS Number: 3037676-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR