1-(2,4-Dichlorothiazol-5-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

456884-5G 456884-1G 456884-500MG
77655-842EA 2417 USD
77655-842 77642-944 77650-908
1-(2,4-Dichlorothiazol-5-yl)-2,2,2-trifluoroethanone ≥97%
1-(2,4-Dichlorothiazol-5-yl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 250.0258 g/mol
MW: C₅Cl₂F₃NOS
Storage Temperature: Ambient
CAS Number: 1823871-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR