2-Bromo-3,5-di-tert-butylphenol ≥97%

Supplier: AOBChem USA

Warning

456872-5G 456872-25G 456872-1G 456872-500MG
77629-456EA 1484.23 USD
77629-456 77655-840 77642-942 77650-906
2-Bromo-3,5-di-tert-butylphenol ≥97%
2-Bromo-3,5-di-tert-butylphenol

Precaución: For Research Use Only

Formula: 285.2199 g/mol
MW: C₁₄H₂₁BrO
Storage Temperature: Ambient
CAS Number: 153872-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR