Benzo[b]thiophene, 2,3-dibromo-, 1,1-dioxide ≥97%

Supplier: AOBChem USA

Warning

454443-5G 454443-25G 454443-1G 454443-500MG
77633-202EA 933 USD
77633-202 77655-828 77642-936 77650-896
Benzo[b]thiophene, 2,3-dibromo-, 1,1-dioxide ≥97%
Benzo[b]thiophene, 2,3-dibromo-, 1,1-dioxide

Precaución: For Research Use Only

Formula: 323.99 g/mol
MW: C₈H₄Br₂O₂S
Storage Temperature: Ambient
CAS Number: 19163-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR