2-Bromo-3-methylbenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

453962-5G 453962-25G 453962-1G 453962-500MG
77633-200EA 856 USD
77633-200 77659-430 77642-934 77650-892
2-Bromo-3-methylbenzo[b]thiophene ≥95%
2-Bromo-3-methylbenzo[b]thiophene

Precaución: For Research Use Only

Formula: 227.12 g/mol
MW: C₉H₇BrS
Storage Temperature: Ambient
CAS Number: 5381-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR