2-Acetyl-3-methylbenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

453957-5G 453957-1G 453957-25G 453957-500MG
77633-198EA 849 USD
77633-198 77638-884 77659-428 77646-908
2-Acetyl-3-methylbenzo[b]thiophene ≥95%
2-Acetyl-3-methylbenzo[b]thiophene

Precaución: For Research Use Only

Formula: C₁₁H₁₀OS
MW: 190.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052499
CAS Number: 18781-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR