1-(2-Chloroethoxy)-4-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

453829-5G 453829-25G 453829-1G 453829-500MG
77633-196EA 1326 USD
77633-196 77659-426 77642-932 77646-906
1-(2-Chloroethoxy)-4-(methylthio)benzene ≥95%
1-(2-Chloroethoxy)-4-(methylthio)benzene

Precaución: For Research Use Only

Formula: 202.7 g/mol
MW: C₉H₁₁ClOS
Storage Temperature: Ambient
CAS Number: 34334-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR