2-(2,3-Difluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

453533-1G 453533-5G 453533-500MG
77638-882EA 605 USD
77638-882 77659-424 77646-904
2-(2,3-Difluorophenyl)propan-2-ol ≥95%
2-(2,3-Difluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 172.172 g/mol
MW: C₉H₁₀F₂O
Storage Temperature: Ambient
CAS Number: 173276-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR