2-(2,6-Difluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

453487-5G 453487-1G 453487-25G 453487-500MG
77633-194EA 1171 USD
77633-194 77638-880 77655-824 77650-890
2-(2,6-Difluorophenyl)propan-2-ol ≥95%
2-(2,6-Difluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 172.172 g/mol
MW: C₉H₁₀F₂O
Storage Temperature: Ambient
CAS Number: 105999-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR