1-(2-Fluoro-4-methyl-3-(methylthio)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

452987-5G 452987-1G 452987-25G 452987-500MG
77629-448EA 1484.23 USD
77629-448 77638-878 77659-422 77650-888
1-(2-Fluoro-4-methyl-3-(methylthio)phenyl)cyclobutanol ≥95%
1-(2-Fluoro-4-methyl-3-(methylthio)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₂H₁₅FOS
Storage Temperature: Ambient
CAS Number: 3037693-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR