1-(2-Fluoro-4-methyl-3-(methylthio)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

452986-5G 452986-1G 452986-25G 452986-500MG
77629-446EA 1484.23 USD
77629-446 77638-876 77655-822 77646-902
1-(2-Fluoro-4-methyl-3-(methylthio)phenyl)cyclopentanol ≥95%
1-(2-Fluoro-4-methyl-3-(methylthio)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 240.337 g/mol
MW: C₁₃H₁₇FOS
Storage Temperature: Ambient
CAS Number: 3037682-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR