2-(2-Fluoro-4-methyl-3-(methylthio)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

452983-5G 452983-1G 452983-25G 452983-500MG
77629-444EA 1484.23 USD
77629-444 77638-874 77659-420 77650-886
2-(2-Fluoro-4-methyl-3-(methylthio)phenyl)propan-2-ol ≥95%
2-(2-Fluoro-4-methyl-3-(methylthio)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 214.3 g/mol
MW: C₁₁H₁₅FOS
Storage Temperature: Ambient
CAS Number: 3037705-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR