1-(4-Chloro-3-ethoxy-2-fluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

452877-5G 452877-1G 452877-500MG 452877-25G
77629-440EA 1777.78 USD
77629-440 77638-870 77652-846 77659-416
1-(4-Chloro-3-ethoxy-2-fluorophenyl)cyclobutanol ≥95%
1-(4-Chloro-3-ethoxy-2-fluorophenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 244.69 g/mol
MW: C₁₂H₁₄ClFO₂
Storage Temperature: Ambient
CAS Number: 3037676-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR