1-(4-Chloro-3-ethoxy-2-fluorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

452876-5G 452876-1G 452876-25G 452876-500MG
77629-438EA 1484.23 USD
77629-438 77638-868 77659-414 77650-882
1-(4-Chloro-3-ethoxy-2-fluorophenyl)cyclopentanol ≥95%
1-(4-Chloro-3-ethoxy-2-fluorophenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 258.716 g/mol
MW: C₁₃H₁₆ClFO₂
Storage Temperature: Ambient
CAS Number: 3037704-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR