2-(4-Chloro-3-ethoxy-2-fluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

452875-5G 452875-500MG 452875-25G 452875-1G
77629-436EA 1484.23 USD
77629-436 77652-844 77659-412 77642-928
2-(4-Chloro-3-ethoxy-2-fluorophenyl)propan-2-ol ≥95%
2-(4-Chloro-3-ethoxy-2-fluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 232.679 g/mol
MW: C₁₁H₁₄ClFO₂
Storage Temperature: Ambient
CAS Number: 3037705-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR