1-(4-Chloro-3-ethoxy-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

452848-5G 452848-1G 452848-25G 452848-500MG
77629-432EA 1484.23 USD
77629-432 77638-864 77659-408 77650-878
1-(4-Chloro-3-ethoxy-2-fluorophenyl)ethanol ≥95%
1-(4-Chloro-3-ethoxy-2-fluorophenyl)ethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₂ClFO₂
Storage Temperature: Ambient
CAS Number: 3037704-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR