(4-Chloro-3-ethoxy-2-fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

452847-5G 452847-25G 452847-1G 452847-500MG
77633-190EA 1441 USD
77633-190 77659-406 77646-898 77650-876
(4-Chloro-3-ethoxy-2-fluorophenyl)trimethylsilane ≥95%
(4-Chloro-3-ethoxy-2-fluorophenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 246.781 g/mol
MW: C₁₁H₁₆ClFOSi
Storage Temperature: Ambient
CAS Number: 3037695-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR