2-(Benzyloxy)-5-fluoropyridin-3-ylboronic acid ≥95%

Supplier: AOBChem USA

Warning

45272-5G 45272-25G 45272-1G 45272-500MG
77629-430EA 1484.23 USD
77629-430 77635-854 77642-926 77650-872
2-(Benzyloxy)-5-fluoropyridin-3-ylboronic acid ≥95%
2-(Benzyloxy)-5-fluoropyridin-3-ylboronic acid

Precaución: For Research Use Only

Formula: 247.03 g/mol
MW: C₁₂H₁₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD18382881
CAS Number: 1206776-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR