(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

45257-5G 45257-25G 45257-1G 45257-500MG
77635-026EA 414 USD
77635-026 77659-402 77642-922 77646-896
(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)boronic acid ≥95%
(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)boronic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₁BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30834357
CAS Number: 2377605-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR