2-(3,5-Bis(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

452206-5G 452206-1G 452206-25G 452206-500MG
77633-182EA 765 USD
77633-182 77638-856 77655-816 77646-894
2-(3,5-Bis(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(3,5-Bis(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 272.187 g/mol
MW: C₁₁H₁₀F₆O
Storage Temperature: Ambient
CAS Number: 67570-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR