1-(4-Fluoro-3,5-dimethylphenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

452194-5G 452194-25G 452194-1G 452194-500MG
77633-180EA 1381 USD
77633-180 77655-814 77642-916 77646-892
1-(4-Fluoro-3,5-dimethylphenyl)-2-methylpropan-1-one ≥95%
1-(4-Fluoro-3,5-dimethylphenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: 194.245 g/mol
MW: C₁₂H₁₅FO
Storage Temperature: Ambient
CAS Number: 1478923-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR