2-(3-Chloro-4-methoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

452186-5G 452186-25G 452186-1G 452186-500MG
77629-422EA 1484.23 USD
77629-422 77655-812 77642-914 77650-862
2-(3-Chloro-4-methoxyphenyl)propan-2-ol ≥95%
2-(3-Chloro-4-methoxyphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₀H₁₃ClO₂
Storage Temperature: Ambient
CAS Number: 850564-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR