4-(Diethoxymethyl)-3-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

452150-1G 452150-5G 452150-500MG
77638-852EA 633 USD
77638-852 77655-808 77650-858
4-(Diethoxymethyl)-3-fluoropyridine ≥97%
4-(Diethoxymethyl)-3-fluoropyridine

Precaución: For Research Use Only

Formula: 199.2220 g/mol
MW: C₁₀H₁₄FNO₂
Storage Temperature: Ambient
CAS Number: 2283558-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR