2-Bromo-6-fluoro-3-iodo-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

452102-5G 452102-25G 452102-1G 452102-500MG
77629-414EA 1484.23 USD
77629-414 77655-806 77646-884 77646-886
2-Bromo-6-fluoro-3-iodo-5-methoxybenzaldehyde ≥97%
2-Bromo-6-fluoro-3-iodo-5-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: 358.931 g/mol
MW: C₈H₅BrFIO₂
Storage Temperature: Ambient
CAS Number: 3031808-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR