3-(2-Methylnaphthalen-1-yl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

449008-5G 449008-500MG 449008-25G 449008-1G
77629-410EA 1484.23 USD
77629-410 77652-836 77655-804 77642-904
3-(2-Methylnaphthalen-1-yl)oxetan-3-ol ≥95%
3-(2-Methylnaphthalen-1-yl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 214.26 g/mol
MW: C₁₄H₁₄O₂
Storage Temperature: Ambient
CAS Number: 3031808-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR