(4-Chloro-2-methylphenyl)(cyclohexyl)methanone ≥95%

Supplier: AOBChem USA

Warning

448991-5G 448991-1G 448991-500MG
77659-388EA 2001 USD
77659-388 77642-902 77646-882
(4-Chloro-2-methylphenyl)(cyclohexyl)methanone ≥95%
(4-Chloro-2-methylphenyl)(cyclohexyl)methanone

Precaución: For Research Use Only

Formula: 236.737 g/mol
MW: C₁₄H₁₇ClO
Storage Temperature: Ambient
CAS Number: 1267629-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR