(4-Fluoronaphthalen-1-yl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

448967-5G 448967-25G 448967-1G 448967-500MG
77633-174EA 708 USD
77633-174 77659-382 77642-896 77646-878
(4-Fluoronaphthalen-1-yl)(methyl)sulfane ≥95%
(4-Fluoronaphthalen-1-yl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 192.253 g/mol
MW: C₁₁H₉FS
Storage Temperature: Ambient
CAS Number: 59080-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR