1-(4-Methoxy-2-(trifluoromethyl)phenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

448944-5G 448944-1G 448944-500MG
77655-798EA 2073 USD
77655-798 77642-894 77650-852
1-(4-Methoxy-2-(trifluoromethyl)phenyl)-2-methylpropan-1-one ≥95%
1-(4-Methoxy-2-(trifluoromethyl)phenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: 246.226 g/mol
MW: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
CAS Number: 2353718-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR