1-(4-Methoxy-2-(trifluoromethyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

448943-5G 448943-25G 448943-1G 448943-500MG
77629-396EA 1484.23 USD
77629-396 77655-796 77642-892 77646-872
1-(4-Methoxy-2-(trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(4-Methoxy-2-(trifluoromethyl)phenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 246.226 g/mol
MW: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
CAS Number: 3048040-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR