(2,4-Bis(trifluoromethyl)phenyl)(cyclopentyl)methanone ≥95%

Supplier: AOBChem USA

Warning

448933-5G 448933-1G 448933-500MG
77659-374EA 2001 USD
77659-374 77642-888 77650-846
(2,4-Bis(trifluoromethyl)phenyl)(cyclopentyl)methanone ≥95%
(2,4-Bis(trifluoromethyl)phenyl)(cyclopentyl)methanone

Precaución: For Research Use Only

Formula: 310.235 g/mol
MW: C₁₄H₁₂F₆O
Storage Temperature: Ambient
CAS Number: 2173122-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR