1-(2,4-Bis(trifluoromethyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

448926-5G 448926-25G 448926-1G 448926-500MG
77629-388EA 1484.23 USD
77629-388 77655-788 77642-882 77646-866
1-(2,4-Bis(trifluoromethyl)phenyl)propan-1-ol ≥95%
1-(2,4-Bis(trifluoromethyl)phenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 272.187 g/mol
MW: C₁₁H₁₀F₆O
Storage Temperature: Ambient
CAS Number: 2410512-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR