1-(3-Chloro-4-(trifluoromethyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

448904-5G 448904-1G 448904-25G 448904-500MG
77633-168EA 1357 USD
77633-168 77638-834 77659-372 77650-842
1-(3-Chloro-4-(trifluoromethyl)phenyl)propan-1-ol ≥95%
1-(3-Chloro-4-(trifluoromethyl)phenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 238.634 g/mol
MW: C₁₀H₁₀ClF₃O
Storage Temperature: Ambient
CAS Number: 2303505-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR