1-(3-Chloro-4-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

448903-5G 448903-1G 448903-500MG
77629-382EA 1484.23 USD
77629-382 77638-832 77650-840
1-(3-Chloro-4-(trifluoromethyl)phenyl)ethanone ≥95%
1-(3-Chloro-4-(trifluoromethyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 222.592 g/mol
MW: C₉H₆ClF₃O
Storage Temperature: Ambient
CAS Number: 129322-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR