(2-(Benzyloxy)phenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

448390-5G 448390-1G 448390-500MG
77659-368EA 2113 USD
77659-368 77642-878 77650-836
(2-(Benzyloxy)phenyl)(cyclopropyl)methanone ≥95%
(2-(Benzyloxy)phenyl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: 252.308 g/mol
MW: C₁₇H₁₆O₂
Storage Temperature: Ambient
CAS Number: 1861670-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR