1-(2-(Benzyloxy)phenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

448389-5G 448389-1G 448389-500MG
77629-380EA 1484.23 USD
77629-380 77646-860 77646-862
1-(2-(Benzyloxy)phenyl)-2-methylpropan-1-one ≥95%
1-(2-(Benzyloxy)phenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: C₁₇H₁₈O₂
Storage Temperature: Ambient
CAS Number: 64686-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR