(2-Bromo-4-ethylphenyl)(thiomorpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

448093-5G 448093-1G 448093-25G 448093-500MG
77633-164EA 1441 USD
77633-164 77638-824 77655-782 77650-832
(2-Bromo-4-ethylphenyl)(thiomorpholino)methanone ≥95%
(2-Bromo-4-ethylphenyl)(thiomorpholino)methanone

Precaución: For Research Use Only

Formula: C₁₃H₁₆BrNOS
Storage Temperature: Ambient
CAS Number: 3031792-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR