Ethyl 2-(4'-methyl-[1,1'-biphenyl]-4-yl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

447644-5G 447644-25G 447644-1G 447644-500MG
77629-376EA 1484.23 USD
77629-376 77635-844 77638-822 77650-830
Ethyl 2-(4'-methyl-[1,1'-biphenyl]-4-yl)-2-oxoacetate ≥97%
Ethyl 2-(4'-methyl-[1,1'-biphenyl]-4-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 268.3071 g/mol
MW: C₁₇H₁₆O₃
Storage Temperature: Ambient
CAS Number: 1267391-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR