2,4-Dichloro-3,6-difluoroaniline ≥95%

Supplier: AOBChem USA

Warning

447637-5G 447637-1G 447637-25G 447637-500MG
77633-162EA 1288 USD
77633-162 77638-818 77655-780 77650-828
2,4-Dichloro-3,6-difluoroaniline ≥95%
2,4-Dichloro-3,6-difluoroaniline

Precaución: For Research Use Only

Formula: 197.9975 g/mol
MW: C₆H₃Cl₂F₂N
Storage Temperature: Ambient
CAS Number: 1314936-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR