4-(1-Hydroxy-2-methylpropyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

447629-5G 447629-25G 447629-1G 447629-500MG
77629-372EA 1484.23 USD
77629-372 77659-364 77642-874 77650-826
4-(1-Hydroxy-2-methylpropyl)benzonitrile ≥97%
4-(1-Hydroxy-2-methylpropyl)benzonitrile

Precaución: For Research Use Only

Formula: 175.2270 g/mol
MW: C₁₁H₁₃NO
Storage Temperature: Ambient
CAS Number: 522653-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR