1-(3-(Ethylthio)phenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

447295-5G 447295-25G 447295-1G 447295-500MG
77629-370EA 1484.23 USD
77629-370 77655-778 77642-872 77650-824
1-(3-(Ethylthio)phenyl)cyclohexanol ≥95%
1-(3-(Ethylthio)phenyl)cyclohexanol

Precaución: For Research Use Only

Formula: C₁₄H₂₀OS
Storage Temperature: Ambient
CAS Number: 3031788-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR