4-(3-(Ethylthio)phenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

447293-5G 447293-25G 447293-1G 447293-500MG
77629-368EA 1821.04 USD
77629-368 77659-362 77642-870 77650-822
4-(3-(Ethylthio)phenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(3-(Ethylthio)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 238.346 g/mol
MW: C₁₃H₁₈O₂S
Storage Temperature: Ambient
CAS Number: 2565807-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR