1-(3-(Ethylthio)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

447290-5G 447290-1G 447290-25G 447290-500MG
77629-366EA 1484.23 USD
77629-366 77638-816 77655-776 77646-856
1-(3-(Ethylthio)phenyl)cyclopentanol ≥95%
1-(3-(Ethylthio)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 222.346 g/mol
MW: C₁₃H₁₈OS
Storage Temperature: Ambient
CAS Number: 3031813-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR