1-(3-(Ethylthio)phenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

447278-5G 447278-1G 447278-500MG
77655-774EA 2188 USD
77655-774 77642-868 77650-820
1-(3-(Ethylthio)phenyl)prop-2-en-1-ol ≥95%
1-(3-(Ethylthio)phenyl)prop-2-en-1-ol

Precaución: For Research Use Only

Formula: 194.293 g/mol
MW: C₁₁H₁₄OS
Storage Temperature: Ambient
CAS Number: 706816-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR