1-(3-(Ethylthio)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

447272-5G 447272-25G 447272-1G 447272-500MG
77629-364EA 1484.23 USD
77629-364 77655-772 77642-866 77646-854
1-(3-(Ethylthio)phenyl)-2-methylpropan-1-ol ≥95%
1-(3-(Ethylthio)phenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 210.336 g/mol
MW: C₁₂H₁₈OS
Storage Temperature: Ambient
CAS Number: 3031799-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR