Ethyl 2-(3-ethoxy-5-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

446930-5G 446930-25G 446930-500MG 446930-1G
77629-360EA 1484.23 USD
77629-360 77635-840 77652-824 77642-862
Ethyl 2-(3-ethoxy-5-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-ethoxy-5-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₃H₁₆O₄
Storage Temperature: Ambient
CAS Number: 3031799-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR